2-Methoxy-3-methyl-5-nitropyridine 98%

Supplier: Ambeed
Warning

A118136-1G A118136-250MG
76810-288EA 11.67 USD
76810-288 76810-290
2-Methoxy-3-methyl-5-nitropyridine 98%
2-Methoxy-3-methyl-5-nitropyridine

Precaución: for Research Use Only

Formula: C₇H₈N₂O₃
MW: 168.1500 g/mol
MDL Number: MFCD04972416
CAS Number: 89694-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR