2-(5-Fluoropyridin-3-yl)acetic acid 95%

Supplier: Ambeed
Warning

A117682-100MG A117682-1G A117682-250MG
76810-274EA 81.69 USD
76810-274 76810-276 76810-278
2-(5-Fluoropyridin-3-yl)acetic acid 95%
2-(5-Fluoropyridin-3-yl)acetic acid

Precaución: for Research Use Only

Formula: C₇H₆FNO₂
MDL Number: MFCD09924977
CAS Number: 38129-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR