(6-Bromo-2-ethoxy-3-fluorophenyl)boronic acid 95%

Supplier: Ambeed
Warning

A117466-1G A117466-250MG
76810-260EA 24 USD
76810-260 76810-262
(6-Bromo-2-ethoxy-3-fluorophenyl)boronic acid 95%
(6-Bromo-2-ethoxy-3-fluorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BBrFO₃
MW: 262.86 g/mol
MDL Number: MFCD12026721
CAS Number: 1310403-98-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR