3-Hydroxy-5-methylbenzonitrile 95%

Supplier: Ambeed
Warning

A117413-1G A117413-250MG A117413-5G
76810-246EA 81 USD
76810-246 76810-248 76810-250
3-Hydroxy-5-methylbenzonitrile 95%
3-Hydroxy-5-methylbenzonitrile

Precaución: for Research Use Only

Formula: C8H7NO
MW: 133.1500 g/mol
MDL Number: MFCD16999137
CAS Number: 95658-81-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR