3-Formyl-4-nitrobenzonitrile 97%

Supplier: Ambeed
Warning

A117232-100MG A117232-10G A117232-1G A117232-250MG A117232-5G
76810-234EA 13 USD
76810-234 76810-236 76810-238 76810-240 77882-961
3-Formyl-4-nitrobenzonitrile 97%
3-Formyl-4-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₈H₄N₂O₃
MDL Number: MFCD10696868
CAS Number: 90178-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR