4'-Hydroxy-3'-nitroacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Hydroxy-3-nitrophenyl)-1-ethanone

A116098-100G A116098-10G A116098-25G A116098-500G
76810-190EA 54 USD
76810-190 76810-192 76810-194 76810-196
4'-Hydroxy-3'-nitroacetophenone 98%
4'-Hydroxy-3'-nitroacetophenone

Precaución: for Research Use Only

Formula: C₈H₇NO₄
MW: 181.15
MDL Number: MFCD00017002
CAS Number: 6322-56-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR