2-(2-Bromophenyl)pyridine 97%

Supplier: Ambeed
Warning

A115899-1G A115899-5G
76810-178EA 150 USD
76810-178 76810-180
2-(2-Bromophenyl)pyridine 97%
2-(2-Bromophenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₈BrN
MDL Number: MFCD15529755
CAS Number: 109306-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR