2-Chloro-3-(trifluoromethoxy)pyridine 95%

Supplier: Ambeed
Warning

A115837-100MG A115837-1G A115837-250MG A115837-5G
76810-166EA 72.15 USD
76810-166 76810-168 76810-170 76810-172
2-Chloro-3-(trifluoromethoxy)pyridine 95%
2-Chloro-3-(trifluoromethoxy)pyridine

Precaución: for Research Use Only

Formula: C₆H₃ClF₃NO
MW: 197.542 g/mol
MDL Number: MFCD17168453
CAS Number: 1206980-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR