Methyl-1-aminocyclopentanecarboxylate hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 1-Aminocyclopentanecarboxylic Acid Methyl Ester Hydrochloride, Methyl 1-aminocyclopentanecarboxylate hydrochloride, Methyl 1-Aminocyclopentanecarboxylate HCl, Cycloleucine Methyl Ester Hydrochloride

A115634-1G A115634-250MG A115634-25G A115634-5G A115634-100G
76810-162EA 8 USD
76810-162 76810-164 77570-940 77570-942 77882-953
Methyl-1-aminocyclopentanecarboxylate hydrochloride 95%
Methyl-1-aminocyclopentanecarboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₃NO₂
MDL Number: MFCD03840364
CAS Number: 60421-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR