2,4-Dihydroxyl-6-fluoroquinazoline 98%

Supplier: Ambeed
Warning

Synonyms: 6-Fluoroquinazoline-2,4-diol

A115537-1G A115537-250MG
76810-148EA 55.17 USD
76810-148 76810-150
2,4-Dihydroxyl-6-fluoroquinazoline 98%
2,4-Dihydroxyl-6-fluoroquinazoline

Precaución: for Research Use Only

Formula: C₈H₅FN₂O₂
MW: 180.14 g/mol
MDL Number: MFCD12024573
CAS Number: 88145-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR