5-Methylindolin-2-one 95%

Supplier: Ambeed
Warning

Synonyms: 5-Methyl-2-oxindole

A115525-100MG A115525-250MG A115525-1G
76810-144EA 7.43 USD
76810-144 76810-146 77670-622
5-Methylindolin-2-one 95%
5-Methylindolin-2-one

Precaución: for Research Use Only

Formula: C₉H₉NO
MW: 147.18 g/mol
MDL Number: MFCD02379906
CAS Number: 3484-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR