6-Chloro-3-formyl-2-picoline 97%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-2-methylpyridine-3-carbaldehyde, 6-Chloro-3-formyl-2-methylpyridine, 6-Chloro-2-methylnicotinaldehyde

A115501-1G A115501-250MG A115501-25G A115501-5G
76810-136EA 98.67 USD
76810-136 76810-138 76810-140 76810-142
6-Chloro-3-formyl-2-picoline 97%
6-Chloro-3-formyl-2-picoline

Precaución: for Research Use Only

Formula: C₇H₆ClNO
MW: 155.5800 g/mol
MDL Number: MFCD07368876
CAS Number: 884495-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR