cis-3-(tert-Butoxycarbonylamino)cyclobutanecarboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A115267-100MG
A115267-250MG
A115267-25G
A115267-5G
A115267-1G
76810-120EA
11
USD
76810-120
76810-122
76810-124
76810-126
77566-060
cis-3-(tert-Butoxycarbonylamino)cyclobutanecarboxylic acid 97%
cis-3-(tert-Butoxycarbonylamino)cyclobutanecarboxylic acid
Precaución: For research use only.
Formula:
C₁₀H₁₇NO₄ |
MDL Number:
MFCD06657680 CAS Number: 1008773-79-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...