Methyl 4,6-dihydroxypyridazine-3-carboxylate 97%

Supplier: Ambeed
Warning

A115085-100MG A115085-250MG A115085-5G
76810-104EA 8 USD
76810-104 76810-106 76810-108
Methyl 4,6-dihydroxypyridazine-3-carboxylate 97%
Methyl 4,6-dihydroxypyridazine-3-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆N₂O₄
MDL Number: MFCD09953611
CAS Number: 372118-00-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR