3-Ethyl-1-phenyl-1H-pyrazol-5-amine 95%

Supplier: Ambeed
Warning

A114465-100MG A114465-1G A114465-250MG
76810-090EA 48.81 USD
76810-090 76810-092 76810-094
3-Ethyl-1-phenyl-1H-pyrazol-5-amine 95%
3-Ethyl-1-phenyl-1H-pyrazol-5-amine

Precaución: for Research Use Only

Formula: C11H13N3
MW: 187.2400 g/mol
MDL Number: MFCD02255835
CAS Number: 1017781-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR