2,4-Difluoro-N-methyl-benzylamine 95%

Supplier: Ambeed
Warning

Synonyms: 1-(2,4-Difluorophenyl)-N-methylmethanamine

A113938-250MG A113938-1G
76810-088EA 34 USD
76810-088 77592-210
2,4-Difluoro-N-methyl-benzylamine 95%
2,4-Difluoro-N-methyl-benzylamine

Precaución: for Research Use Only

Formula: C₈H₉F₂N
MDL Number: MFCD09047006
CAS Number: 696589-32-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR