3,4-Dimethoxybenzonitrile 98%

Supplier: Ambeed
Danger

A113887-25G A113887-5G
76810-080EA 21 USD
76810-080 77570-754
3,4-Dimethoxybenzonitrile 98%
3,4-Dimethoxybenzonitrile

Precaución: for Research Use Only

Formula: C₉H₉NO₂
MDL Number: MFCD00001802
CAS Number: 2024-83-1
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR