(3,4-Dichlorophenyl)acetic acid methyl ester 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3,4-Dichlorophenylacetate, 3,4-Dichlorophenylacetic acid methyll ester, Methyl 2-(3,4-dichlorophenyl)acetate, Methyl (3,4-dichlorophenyl)acetate

A113689-10G A113689-1G A113689-25G A113689-5G
76810-066EA 11.67 USD
76810-066 76810-068 76810-070 76810-072
(3,4-Dichlorophenyl)acetic acid methyl ester 98%
(3,4-Dichlorophenyl)acetic acid methyl ester

Precaución: for Research Use Only

Formula: (C₆H₃Cl₂)CH₂COOCH₃
MW: 219.07 g/mol
MDL Number: MFCD00800680
CAS Number: 6725-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR