(S)-2-(((Benzyloxy)carbonyl)amino)pent-4-enoic acid 98%

Supplier: Ambeed
Warning

A113544-100MG A113544-1G A113544-250MG A113544-5G
76810-056EA 8.49 USD
76810-056 76810-058 76810-060 76810-062
(S)-2-(((Benzyloxy)carbonyl)amino)pent-4-enoic acid 98%
(S)-2-(((Benzyloxy)carbonyl)amino)pent-4-enoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO₄
MW: 249.26 g/mol
MDL Number: MFCD02094548
CAS Number: 78553-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR