4'-Bromo-3'-chloroacetophenone 97%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Bromo-3-chlorophenyl)ethanone

A113506-10G A113506-1G A113506-25G A113506-5G
76810-048EA 80.63 USD
76810-048 76810-050 76810-052 76810-054
4'-Bromo-3'-chloroacetophenone 97%
4'-Bromo-3'-chloroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆BrClO
MW: 233.4900 g/mol
MDL Number: MFCD14636445
CAS Number: 3114-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR