(cis-2-((1H-Imidazol-1-yl)methyl)-2-(2,4-dichlorophenyl)-1,3-dioxolan-4-yl)methyl 4-methylbenzenesulfonate 95%

Supplier: Ambeed
Warning

Synonyms: trans-Ketoconazole EP Impurity E

A112944-100G A112944-10G A112944-1G A112944-25G A112944-5G
76810-020EA 109.28 USD
76810-020 76810-022 76810-024 76810-026 76810-028
(cis-2-((1H-Imidazol-1-yl)methyl)-2-(2,4-dichlorophenyl)-1,3-dioxolan-4-yl)methyl 4-methylbenzenesulfonate 95%
(cis-2-((1H-Imidazol-1-yl)methyl)-2-(2,4-dichlorophenyl)-1,3-dioxolan-4-yl)methyl 4-methylbenzenesulfonate

Precaución: for Research Use Only

Formula: C₂₁H₂₀Cl₂N₂O₅S
MW: 483.37 g/mol
MDL Number: MFCD07367827
CAS Number: 134071-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR