3-Chloro-4-iodobenzotrifluoride 95% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilized with Copper chip

A112874-1G A112874-25G A112874-5G
76810-014EA 9.55 USD
76810-014 76810-016 76810-018
3-Chloro-4-iodobenzotrifluoride 95% stabilized
3-Chloro-4-iodobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₃ClF₃I
MW: 306.45 g/mol
MDL Number: MFCD00042562
CAS Number: 141738-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR