2-Chloro-5-hydroxybenzaldehyde 98%

Supplier: Ambeed
Warning

A112558-10G A112558-1G A112558-250MG
76809-994EA 545.31 USD
76809-994 76809-996 76809-998
2-Chloro-5-hydroxybenzaldehyde 98%
2-Chloro-5-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₅ClO₂
MW: 156.57 g/mol
MDL Number: MFCD07783652
CAS Number: 7310-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR