2-(3-Methoxy-4-nitrophenyl)acetic acid 97%

Supplier: Ambeed
Warning

A110994-1G A110994-250MG A110994-5G A110994-100MG
76809-946EA 147 USD
76809-946 76809-948 76809-950 77882-929
2-(3-Methoxy-4-nitrophenyl)acetic acid 97%
2-(3-Methoxy-4-nitrophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₉NO₅
MDL Number: MFCD09997710
CAS Number: 5803-22-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR