Febantel 98+%

Supplier: Ambeed
Warning

Synonyms: N-(((2-(2-MEthoxyacetamido)-4-(phenylsulphanyl)phenyl)amino)((methoxycarbonyl)imino)methyl)carbamate

A110936-100MG A110936-1G A110936-25G A110936-5G A110936-100G
76809-932EA 5.31 USD
76809-932 76809-934 76809-936 76809-938 77670-572
Febantel 98+%
Febantel

Precaución: for Research Use Only

Formula: C₂₀H₂₂N₄O₆S
MW: 446.48 g/mol
MDL Number: MFCD01738527
CAS Number: 58306-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR