2-Methoxy-4-(methylsulfonyl)aniline 98%

Supplier: Ambeed
Warning

A110846-1G A110846-5G A110846-100MG A110846-250MG
76809-922EA 82.76 USD
76809-922 76809-924 77610-876 77610-878
2-Methoxy-4-(methylsulfonyl)aniline 98%
2-Methoxy-4-(methylsulfonyl)aniline

Precaución: for Research Use Only

Formula: C₈H₁₁NO₃S
MW: 201.24 g/mol
MDL Number: MFCD18861008
CAS Number: 41608-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR