4-Bromo-2-methylbenzyl bromide 97%

Supplier: Ambeed
Danger

Synonyms: 5-Bromo-2-(bromomethyl)toluene, 4-Bromo-1-(bromomethyl)-2-methylbenzene

A110168-100MG A110168-1G A110168-250MG
76809-896EA 60.48 USD
76809-896 76809-898 76809-900
4-Bromo-2-methylbenzyl bromide 97%
4-Bromo-2-methylbenzyl bromide

Precaución: for Research Use Only

Formula: C₈H₈Br₂
MW: 263.957 g/mol
MDL Number: MFCD12025011
CAS Number: 156001-49-9
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR