6-Chloroimidazo[1,2-a]pyridine-3-carbaldehyde 97%

Supplier: Ambeed
Warning

A110156-250MG A110156-1G A110156-5G
76809-894EA 32 USD
76809-894 77670-562 77882-923
6-Chloroimidazo[1,2-a]pyridine-3-carbaldehyde 97%
6-Chloroimidazo[1,2-a]pyridine-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₅ClN₂O
MDL Number: MFCD09909649
CAS Number: 29096-59-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR