tert-Butyl (piperidin-3-ylmethyl)carbamate hydrochloride 97%

Supplier: Ambeed
Warning

A110056-100MG A110056-1G A110056-250MG
76809-888EA 9.55 USD
76809-888 76809-890 76809-892
tert-Butyl (piperidin-3-ylmethyl)carbamate hydrochloride 97%
tert-Butyl (piperidin-3-ylmethyl)carbamate hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₂₃ClN₂O₂
MW: 250.77 g/mol
MDL Number: MFCD11101360
CAS Number: 1159826-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR