7-Iodo-2H-benzo[b][1,4]oxazin-3(4H)-one 95%

Supplier: Ambeed
Warning

A110037-100MG A110037-1G A110037-250MG
76809-882EA 66 USD
76809-882 76809-884 76809-886
7-Iodo-2H-benzo[b][1,4]oxazin-3(4H)-one 95%
7-Iodo-2H-benzo[b][1,4]oxazin-3(4H)-one

Precaución: for Research Use Only

Formula: C₈H₆INO₂
MDL Number: MFCD07774202
CAS Number: 874840-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR