Methyl-5-bromo-3-hydroxypicolinate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-bromo-3-hydroxypicolinate

A109964-100MG A109964-250MG A109964-1G
76809-876EA 77.45 USD
76809-876 76809-878 77565-938
Methyl-5-bromo-3-hydroxypicolinate 97%
Methyl 5-bromo-3-hydroxypicolinate

Precaución: For research use only.

Formula: C₇H₆BrNO₃
MW: 232.0300 g/mol
MDL Number: MFCD12407822
CAS Number: 1242320-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR