4-Propyl-4'-vinyl-1,1'-bi(cyclohexane) 97%

Supplier: Ambeed
Warning

A109728-1G A109728-250MG
76809-868EA 20.16 USD
76809-868 76809-870
4-Propyl-4'-vinyl-1,1'-bi(cyclohexane) 97%
4-Propyl-4'-vinyl-1,1'-bi(cyclohexane)

Precaución: for Research Use Only

Formula: C₁₇H₃₀
MW: 234.42 g/mol
MDL Number: MFCD12911824
CAS Number: 477557-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR