1-(4-Cyanophenyl)piperazine 98%
Supplier: Ambeed
|
Warning
|
A109529-5G
A109529-1G
A109529-25G
76809-858EA
10.61
USD
76809-858
77570-544
77882-919
77882-920
1-(4-Cyanophenyl)piperazine 98%
1-(4-Cyanophenyl)piperazine
Precaución: for Research Use Only
Formula:
C₁₁H₁₃N₃ |
MDL Number:
MFCD01320890 CAS Number: 68104-63-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...