Ethyl 4-bromo-1-methyl-1H-pyrazole-3-carboxylate 97%

Supplier: Ambeed
Warning

A109047-100MG A109047-1G A109047-250MG
76809-830EA 22.28 USD
76809-830 76809-832 76809-834
Ethyl 4-bromo-1-methyl-1H-pyrazole-3-carboxylate 97%
Ethyl 4-bromo-1-methyl-1H-pyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉BrN₂O₂
MW: 233.06 g/mol
MDL Number: MFCD02228182
CAS Number: 400877-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR