4-Bromo-2-ethoxy-1-methylbenzene 95%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2-ethoxytolune

A109038-250MG A109038-5G
76809-826EA 23.34 USD
76809-826 76809-828
4-Bromo-2-ethoxy-1-methylbenzene 95%
4-Bromo-2-ethoxy-1-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁BrO
MW: 215.09 g/mol
MDL Number: MFCD11617247
CAS Number: 871888-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR