6-Methyluracil 98%

Supplier: Ambeed
Warning

Synonyms: 6-Methylpyrimidine-2,4(1H,3H)-dione, 4-Methyluracil, 6-Methyl-2,4(1H,3H)-pyrimidinedione

A108584-100G A108584-25G A108584-500G A108584-1000G
76809-818EA 8.49 USD
76809-818 76809-820 76809-822 77610-856
6-Methyluracil 98%
6-Methyluracil

Precaución: for Research Use Only

Formula: C₅H₆N₂O₂
MW: 126.11 g/mol
MDL Number: MFCD00006028
CAS Number: 626-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR