6-Chloro-5-(trifluoromethyl)pyridin-3-amine 95%
Supplier: Ambeed
|
Warning
|
A107985-100MG
A107985-1G
A107985-250MG
A107985-5G
76809-788EA
41.38
USD
76809-788
76809-790
76809-792
76809-794
6-Chloro-5-(trifluoromethyl)pyridin-3-amine 95%
6-Chloro-5-(trifluoromethyl)pyridin-3-amine
Precaución: for Research Use Only
Formula:
C₆H₄ClF₃N₂ MW: 196.558 g/mol |
MDL Number:
MFCD11101042 CAS Number: 99368-68-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...