Methyl 3-formyl-2-nitrobenzoate 95%

Supplier: Ambeed
Warning

A107781-1G A107781-250MG A107781-25G A107781-5G
76809-772EA 6.37 USD
76809-772 76809-774 76809-776 76809-778
Methyl 3-formyl-2-nitrobenzoate 95%
Methyl 3-formyl-2-nitrobenzoate

Precaución: for Research Use Only

Formula: C₉H₇NO₅
MW: 209.156 g/mol
MDL Number: MFCD14706049
CAS Number: 138229-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR