2-Aminoisobutyric acid 98%

Supplier: Ambeed
Warning

Synonyms: α-Aminoisobutyric acid, 2-Amino-2-methylpropionic acid, 2-Metilalanina, H-Aib-OH, 2-Methylalanine, 2-Amino-2-methylpropanoic acid

A107418-1000G A107418-100G A107418-25G A107418-500G
76809-752EA 89.12 USD
76809-752 76809-754 76809-756 76809-758
2-Aminoisobutyric acid 98%
2-Aminoisobutyric acid

Precaución: for Research Use Only

Formula: C₄H₉NO₂
MW: 103.12 g/mol
MDL Number: MFCD00008049
CAS Number: 62-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR