4-(Ethoxycarbonyl)-2-nitrophenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Ethoxycarbonyl-2-nitrophenylboronic acid, 4-Ethoxycarbonyl-2-nitrobenzeneboronic acid

A107311-5G A107311-25G
76809-748EA 20 USD
76809-748 77880-868
4-(Ethoxycarbonyl)-2-nitrophenylboronic acid 97%
4-(Ethoxycarbonyl)-2-nitrophenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BNO₆
MDL Number: MFCD02179460
CAS Number: 5785-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR