1,2,3,4-Tetrahydroquinoline 98%

Supplier: Ambeed
Warning

A107215-100G A107215-10G A107215-25G A107215-500G A107215-1000G
76809-736EA 19.1 USD
76809-736 76809-738 76809-740 76809-742 77670-532
1,2,3,4-Tetrahydroquinoline 98%
1,2,3,4-Tetrahydroquinoline

Precaución: for Research Use Only

Formula: C₉H₁₁N
MW: 133.19 g/mol
MDL Number: MFCD00006693
CAS Number: 635-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR