1,4-Dihydro-4-pyridinone 97%

Supplier: Ambeed
Warning

Synonyms: 4-HYDROXYPYRIDINE (4-PYRIDINOL), 4(1H)-Pyridone, pyridin-4(1H)-one

A107171-100G A107171-10G A107171-25G A107171-5G
76809-718EA 88.06 USD
76809-718 76809-720 76809-722 76809-724
1,4-Dihydro-4-pyridinone 97%
1,4-Dihydro-4-pyridinone

Precaución: for Research Use Only

Formula: C₅H₅NO
MW: 95.1 g/mol
MDL Number: MFCD00040458
CAS Number: 108-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR