(6-Methoxy-3-pyridyl)methanol 98%

Supplier: Ambeed
Warning

Synonyms: 5-(Hydroxymethyl)-2-methoxypyridine, (6-Methoxypyridin-3-yl)methanol

A107111-1G A107111-250MG A107111-25G A107111-5G
76809-700EA 26.53 USD
76809-700 76809-702 76809-704 76809-706
(6-Methoxy-3-pyridyl)methanol 98%
(6-Methoxy-3-pyridyl)methanol

Precaución: for Research Use Only

Formula: C₇H₉NO₂
MW: 139.16 g/mol
MDL Number: MFCD06202826
CAS Number: 58584-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR