3-Hydroxymethyl-4-methoxybenzeneboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Hydroxymethyl)-4-methoxybenzeneboronic acid, 3-(Hydroxymethyl)-4-methoxyphenylboronic acid, (3-(Hydroxymethyl)-4-methoxyphenyl)boronic acid, 3-Hydroxymethyl-4-methoxyphenylboronic acid

A106928-10G A106928-1G A106928-25G A106928-5G
76809-692EA 101.85 USD
76809-692 76809-694 76809-696 76809-698
3-Hydroxymethyl-4-methoxybenzeneboronic acid 97%
3-Hydroxymethyl-4-methoxybenzeneboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₄
MW: 181.982 g/mol
MDL Number: MFCD14155851
CAS Number: 908142-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR