Boc-(4-carboxymethyl)piperazine 98%

Supplier: Ambeed
Warning

Synonyms: [1-(tert-Butoxycarbonyl)piperazin-4-yl]acetic acid, 2-(4-(tert-Butoxycarbonyl)piperazin-1-yl)acetic acid

A106857-100G A106857-100MG A106857-10G A106857-1G A106857-250MG A106857-25G A106857-5G
76809-678EA 642.91 USD
76809-678 76809-680 76809-682 76809-684 76809-686 76809-688 76809-690
Boc-(4-carboxymethyl)piperazine 98%
Boc-(4-carboxymethyl)piperazine

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₄
MW: 244.29 g/mol
MDL Number: MFCD02682402
CAS Number: 156478-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR