(2-Fluoro-5-(trifluoromethyl)phenyl)acetic acid 98%

Supplier: Ambeed
Warning

A106564-1G A106564-25G A106564-5G
76809-664EA 13 USD
76809-664 76809-666 76809-668 77670-526
(2-Fluoro-5-(trifluoromethyl)phenyl)acetic acid 98%
(2-Fluoro-5-(trifluoromethyl)phenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₆F₄O₂
MDL Number: MFCD00061187
CAS Number: 220227-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR