1-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl)ethanone 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Acetylthiophene-2-boronic acid pinacol ester
A106507-100MG
A106507-1G
A106507-250MG
A106507-5G
76809-654EA
15.92
USD
76809-654
76809-656
76809-658
76809-660
1-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl)ethanone 98+%
1-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl)ethanone
Precaución: for Research Use Only
Formula:
C₁₂H₁₇BO₃S MW: 252.14 g/mol |
MDL Number:
MFCD08063138 CAS Number: 942070-32-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...