N-(tert-Butoxycarbonyl)-D-aspartic acid 97%

Supplier: Ambeed
Warning

Synonyms: N-Boc-D-aspartic Acid, Boc-D-Asp-OH

A105555-1G A105555-5G A105555-100G A105555-25G
76809-594EA 6.37 USD
76809-594 76809-596 77570-328 77570-330
N-(tert-Butoxycarbonyl)-D-aspartic acid 97%
N-(tert-Butoxycarbonyl)-D-aspartic acid

Precaución: for Research Use Only

Formula: C₉H₁₅NO₆
MW: 233.22 g/mol
MDL Number: MFCD00798618
CAS Number: 62396-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR