6-Chloro-4-nitro-1H-indazole 95%

Supplier: Ambeed
Warning

A105534-1G A105534-250MG A105534-5G
76809-588EA 237.65 USD
76809-588 76809-590 76809-592
6-Chloro-4-nitro-1H-indazole 95%
6-Chloro-4-nitro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄ClN₃O₂
MW: 197.579 g/mol
MDL Number: MFCD07781380
CAS Number: 885519-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR