BAPTA tetramethyl ester 98%

Supplier: Ambeed
Warning

A105515-100G A105515-1G A105515-25G A105515-5G
76809-580EA 998 USD
76809-580 76809-582 76809-584 76809-586
BAPTA tetramethyl ester 98%
BAPTA tetramethyl ester

Precaución: for Research Use Only

Formula: C₂₆H₃₂N₂O₁₀
MDL Number: MFCD00532664
CAS Number: 125367-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR